Learn More
Reitenspiess™ Test Tube Brush with Wool Tip
Product Code. 10510202
Change view
Click to view available options
Length (Metric):
260 mm
270 mm
280 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10510202 260 mm Each
10692931 280 mm Each
10097900 270 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10510202 Supplier Reitenspiess™ Supplier No. 70100234
  • ONLINE EXCLUSIVE PRICE
    € 12.74
    € 7.12 / Pack of 1
    Save € 5.62
    44% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Clean your test tubes using the Reitenspiess™ Test Tube Brush with Wool Tip. Use a test tube brush to clean your test tubes and other laboratory ware.
  • Natural bristles
  • Wool Tip
  • Sterile
Specifications
Product Type Test Tube Brush
Sterility Sterile
For Use With (Application) For cleaning, disinfection, sterilization.
Length (Metric) 260 mm
Quantity 1 Ea.