missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natural Killer Cell Antibody (H25), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50526AF647
This item is not returnable.
View return policy
Description
Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
Specifications
| Natural Killer Cell | |
| Monoclonal | |
| Alexa Fluor 647 | |
| NK Cell | |
| HSE-2 cell line | |
| 0.1 mL | |
| Immunology | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| H25 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction