Learn More
Kautex™ LDPE Wide Mouth Bottles Series 303
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
250 mL
300 mL
500 mL
750 mL
1000 mL
1500 mL
2000 mL
Neck ID (Metric):
23.8 mm
30.8 mm
39.2 mm
50 mm
Unit Size:
Each
Pack of 10
Pack of 180
Pack of 20
Pack of 279
Pack of 4
Pack of 5
Pack of 50
Pack of 90
Description
- Manufactured from low density polyethylene (LDPE) for flexibility and toughness
- Natural translucent color, round bottles
- Easy to fill and clean
- Ideal for dispensing, sending samples, adhesives, transportation and storage
- Includes leakproof screw closure
Specifications
Specifications
| Capacity (Metric) | 200 mL |
| For Use With (Application) | Dispensing, Sending Samples, Mailing, Transport, Storage |
| Form | Round |
| Includes | Closure |
| Material | LDPE |
| Neck ID (Metric) | 30.8 mm |
| Replacement Closure | Screw Closure LDPE |
| Quantity | 1 Ea. |
| Graduated | No |
| Height (Metric) | 114 mm |
| Show More |