missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain Natural Rubber GSR Standard Tubing

Use this tubing for a variety of industrial, construction and chemical applications. Saint Gobain Performance Plastics™ Natural Rubber GSR Standard Tubing provides an ideal combination of chemical resistance and pump life.

Brand:  Saint-Gobain 710082

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11761736

  • € 87.60 / 25m


Estimated Shipment
17-02-2021
Add to basket

Description

Description

  • Very good elasticity and flexibility
  • Good resistance to abrasion
  • Meets FDA requirements
Specifications

Specifications

FDA compliant, food WCR
70°C
Natural Rubber
25 m
Handle many industrial applications
Red
6 mm
10 mm
2 mm
-40°C
Videos
SDS
Documents