Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Provides flexibility, tear strength and resilience for vacuum applications. Deutsch Neumann™ Vacuum Quality Laboratory Tubing is manufactured from red natural rubber.
Brand: Deutsch Neumann™ 3020820
Additional Details : Weight : 7.00200kg
Description
- Durometer 50 Shore A
- Good elastic memory
- Excellent choice for vacuum applications
Specifications
8 mm | |
20 mm | |
6 mm |
Natural Rubber | |
25 m | |
Red |