Learn More
Deutsch Neumann™ Vacuum Quality Laboratory Tubing
Provides flexibility, tear strength and resilience for vacuum applications. Deutsch Neumann™ Vacuum Quality Laboratory Tubing is manufactured from red natural rubber.
Brand: Deutsch Neumann™ 3020515
Additional Details : Weight : 3.16800kg
Description
- Durometer 50 Shore A
- Good elastic memory
- Excellent choice for vacuum applications
Specifications
5 mm | |
15 mm | |
5 mm |
Natural Rubber | |
25 m | |
Red |