Learn More
mb2 Stool on Castors - Wood - Mid Lift
Product Code. 11947654
Click to view available options
:
Holds 5 Wheels
Product Code. 11947654
Brand: mb2 AB4B2D1NE2
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Stool on casters - wood seat - Mid lift
  • Stool with wood seat
  • Pneumatic cylinder for height adjustment
  • 5-star reinforced nylon base 640 mm dia.
  • Hard floor castors 50 mm
Specifications
Product Type Stool
Color Natural
Adjustable (Height) Yes
Adjustable Height
Autoclavable Not Autoclavable
Finish Fireproof
Height (Metric) 646 mm
Holds 5 Wheels
Seating Height 470 to 650 mm
Weight (Metric) 5.98 kg
With Armrest No
With Backrest No
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt No
Material Plywood
Seating Type Stool