Learn More
Abeba™ Nature 4080 Shoes
Product Code. 15621438
Click to view available options
Size:
34
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. 15621438
Brand: Abeba™ 4080/44
Leather covered cork footbed is ESD dissipative.
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • White
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • 2 adjustable instep straps
  • velcro fastening
Specifications
Certifications/Compliance CE EN ISO 20347:2012 OB A, FO SRC
Color White
Footbed Material Cork
Material Leather
Size 44
Sole Features Slip-resistant
Sole Material Rubber
Style 4080