Learn More
Novus Biologicals™ ABCB5 Antibody (Brigham and Women's patent anti-ABCB5) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28518-1mg
 View more versions of this product
Product Code. 30047230
  • € 920.00 / 1mg
Description
ABCB5 Monoclonal antibody specifically detects ABCB5 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABCB5
Monoclonal
Unconjugated
Q2M3G0
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Brigham and Women's patent anti-ABCB5
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
ABCB5 P-gp, ABCB5alpha, ABCB5beta, ATP-binding cassette protein, ATP-binding cassette sub-family B member 5, ATP-binding cassette, sub-family B (MDR/TAP), member 5, EC 3.6.3, EC 3.6.3.44, EST422562, P-glycoprotein ABCB5
ABCB5
1 mg
ABC Transporters, Lipid and Metabolism
340273