missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAV1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15713641 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15713641 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15713641 Supplier Invitrogen™ Supplier No. PA555718

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AKSFVKPPSL ANLDKVNSNS LDLPSSSDTT HASKVPDLHA TSSASGGPLP SCFTPSPAPI LNINSASFSQ GLELMSGFSV PKETRMYPKL SGLHRSMESL QMPMSLPSAF PSSTPVPTPP APPAAPTEEE TEELTWSGSP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

NAV1 belongs to the neuron navigator family and is expressed predominantly in the nervous system. The encoded protein contains coiled-coil domains and a conserved AAA domain characteristic for ATPases associated with a variety of cellular activities. This gene is similar to unc-53, a Caenorhabditis elegans gene involved in axon guidance. The exact function of this gene is not known, but it is thought to play a role in in neuronal development and regeneration. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAV1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nav1
Gene Accession No. Q8NEY1
Gene Alias 9530089B19; 9930003A20Rik; C230080M11Rik; KIAA1151; KIAA1213; LOC685707; mKIAA1151; Nav1; Neuron navigator 1; neuron navigator-1; POMFIL3; pore membrane and/or filament interacting like protein 3; pore membrane and/or filament-interacting-like protein 3; similar to neuron navigator 1; STEERIN1; steerin-1; unc-53 homolog 1; Unc53h1
Gene Symbols Nav1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAV1. Recombinant protein control fragment (Product #RP-90900).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89796
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.