Learn More
NAV3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-82951
 View more versions of this product
Product Code. 18790414
  • € 539.00 / 0.10mL
Description
NAV3 Polyclonal specifically detects NAV3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
RUO
IgG
Rabbit
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.