missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NAV3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556931
This item is not returnable.
View return policy
Description
Immunogen sequence: STDDLNTTSS VSSYSNITVP SRKNTQLRTD SEKRSTTDET WDSPEELKKP EEDFDSHGDA GGKWKTVSSG LPEDPEKAGQ KASLS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
Neuron navigator 3 (NAV3), also known as pore membrane and/or filamentinteracting-like protein 1 (POMFIL1), unc-53 homolog 3 (unc53H3) or STEERIN3, is a 2,385 amino acid protein that localizes to the nuclear outer membrane and plays a role in neuron regeneration. Existing as three alternatively spliced isoforms, Neuron navigator 3 is highly expressed in adult and fetal brain and is found at moderate to low levels in ovary, lung, testis, placenta and heart. Neuron navigator 3 is thought be involved in the regulation of IL-2 production and contains one CH (calponin-homology) domain, three N-terminal hydrophobic domains, multiple N-glycosylation sites, three ATP/GTP-binding A motifs (P loops) and several phosphorylation sites.
Specifications
| NAV3 | |
| Polyclonal | |
| Unconjugated | |
| NAV3 | |
| 4732483H20Rik; 9630020C08Rik; KIAA0938; mKIAA0938; NAV3; Neuron navigator 3; POMFIL1; Pomfil1p; pore membrane and/or filament interacting like protein 1; pore membrane and/or filament-interacting-like protein 1; RGD1306259; steerin 3; STEERIN3; Steerin-3; Unc-53 homolog 3; unc53H3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 89795 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IVL0 | |
| NAV3 | |
| Recombinant protein corresponding to Human NAV3. Recombinant protein control fragment (Product #RP-94970). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction