missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NBL1 Monoclonal Antibody (7F8A9D4), Invitrogen™

Mouse Monoclonal Antibody
€ 265.00 - € 461.00
Tekniske data
| Antigen | NBL1 |
|---|---|
| Clone | 7F8A9D4 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
17749863
|
Invitrogen™
MA543516 |
50 μL |
€ 265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17749533
![]() |
Invitrogen™
MA542979 |
100 μL |
€ 461.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
Specificity: Human NBL1/DAN/DAND1
This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.Tekniske data
| NBL1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| D1S1733E; D4H1S1733E; DAN; DAN domain family member 1; Dana; DAND1; differential screening-selected gene aberrant in neuroblastoma; N03; NB; Nbl1; neuroblastoma 1, DAN family BMP antagonist; neuroblastoma candidate region, suppression of tumorigenicity 1; neuroblastoma suppressor of tumorigenicity 1; neuroblastoma, suppression of tumorigenicity 1; Neuroblastoma, suppression of tumorigenicity 1 (DNA segment human D1S1733E); NO3; Protein N03; Zinc finger protein DAN | |
| Nbl1 | |
| IgG2b | |
| Protein A | |
| Antibody |
| 7F8A9D4 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| P41271 | |
| 4681 | |
| Recombinant Human NBL1 protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Nbl1 |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel