Learn More
NBN Mouse anti-Human, Clone: OTI3C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF801448
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.Specifications
| NBN | |
| Monoclonal | |
| Unconjugated | |
| NBN | |
| AT-V1, AT-V2, ATV, NBS, NBS1, P95 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4683 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O60934 | |
| NBN | |
| Human recombit protein fragment corresponding to amino acids 183-460 of human NBN produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.