missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45RB Antibody (PTPRC/2877R), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08924H
This item is not returnable.
View return policy
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Human, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CD45RB | |
| Monoclonal | |
| HRP | |
| B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Recombinant human full-length CD45RB protein (Uniprot: P08575) | |
| 100 μg | |
| Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 5788 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PTPRC/2877R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction