missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NBPF6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103962
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NBPF6.
Members of the neuroblastoma breakpoint family are encoded by genes that map to a region of segmental duplications on human chromosome 1. Both NBPF4 (neuroblastoma breakpoint family member 4) and NBPF6 (neuroblastoma breakpoint family member 6) are cytoplasmic proteins that belong to the NBPF family. NBPF4 contains 123 amino acids and is expressed in testis, while NBPF6 consists of 638 amino acids and contains 3 NBPF domains. Like other NBPF family members, both proteins are encoded by genes that map to human chromosome 1p13.3. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson's disease, Gaucher disease, schizophrenia and Usher syndrome.
Specifications
| NBPF6 | |
| Polyclonal | |
| Unconjugated | |
| NBPF6 | |
| NBPF6; Neuroblastoma breakpoint family member 6; neuroblastoma breakpoint family, member 6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 653149 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5VWK0 | |
| NBPF6 | |
| A synthesized peptide derived from human NBPF6(Accession Q5VWK0), corresponding to amino acid residues E505-L555. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction