missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM-1/CD56 Antibody (CL12732), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18543-25ul
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12732 | |
| Immunohistochemistry 1:5000 - 1:10000, Immunohistochemistry-Paraffin 1:5000 - 1:10000 | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| This antibody was developed against a sythetic peptide for P13591. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 4684 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction