missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM-1/CD56 Antibody (NCAM1/795), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47826APCCY7
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| NCAM1 | |
| Recombinant human NCAM1 protein | |
| 0.1 mL | |
| Primary | |
| This monoclonal antibody reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is also expressed on some mesodermally derived tumors (rhabdomyosarcoma). Anti-CD56 plays an important role in the diagnosis of nodal and nasal NK/T-cell lymphomas. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| NCAM1/795 | |
| Flow Cytometry | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Mouse | |
| Protein A or G purified | |
| Astrocyte Markers, Cellular Markers, Cytokine Research, Cytoskeleton Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| 4684.0 | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction