missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM-1/CD56 Antibody (rNCAM1/8655), Novus Biologicals™
Mouse Monoclonal Antibody
€ 249.00 - € 494.00
Specifications
| Antigen | NCAM-1/CD56 |
|---|---|
| Clone | rNCAM1/8655 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614515
|
Novus Biologicals
NBP3-20517-20ug |
20 μg |
€ 249.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18664364
|
Novus Biologicals
NBP3-20517-100ug |
100 μg |
€ 494.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| NCAM-1/CD56 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 4684 | |
| IgG1 κ | |
| Protein A or G purified |
| rNCAM1/8655 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Recombinant human NCAM-1/CD56 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title