Learn More
NCAM-1/CD56 Antibody (SPM489) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-44714
Product Code. 18620726
  • € 498.00 / 7mL
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCAM-1/CD56
Monoclonal
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin)
SPM489
Immunohistochemistry, Immunohistochemistry-Paraffin 1 - 2 μg/mL
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Membrane preparation of a small cell lung carcinoma
7 mL
Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
4684
Store at 4°C.
IgG1 κ