missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1 Monoclonal Antibody (OTI3H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805182S
This item is not returnable.
View return policy
Description
NCAM1 Monoclonal antibody specifically detects NCAM1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NCAM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q969V6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57591 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H2 | |
| Unconjugated | |
| Mrtfa | |
| adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; Cd56; CD-56; CD56 120 kDa GPI-linked isoform; CD56 140 kDa isoform; CD56 140 kDa VASE isoform; E NCAM; E-NCAM; MSK39; N CAM1; NCAM; N-CAM; Ncam1; N-CAM-1; NCAM-1; NCAMC; NCAM-C; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM | |
| Human recombinant protein fragment corresponding to amino acids 165-397 of human MKL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction