Learn More
NCAM1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1G4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF506208
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD56 (NCAM, neural cell adhesion molecule) is a transmembrane glycoprotein of the immunoglobulin family that serves as an adhesive molecule and is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa. CD56 is found on T cells and NK cells, and is involved in cell migration, axonal growth, pathfinding and synaptic plasticity. Polysialic modification results in reduction of CD56-mediated cell adhesion. Through its extracellular region, CD56 mediates homophilic and heterophilic interactions by binding extracellular matrix components such as laminin and integrins. CD56 is expressed on most neuroectodermal derived cell lines, tissues and neoplasms such as retinoblastoma, medulloblastoma, astrocytomas and neuroblastoma. Further, CD56 is a widely used neuroendocrine marker with a high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms.Specifications
| NCAM1 | |
| Monoclonal | |
| Unconjugated | |
| NCAM1 | |
| CD56, MSK39, NCAM | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 17967, 24586, 4684, 479435 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P13591, P13595, P13596 | |
| NCAM1 | |
| Human recombit protein fragment corresponding to amino acids 20-718 of human NCAM1 produced in HEK293T cells. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.