missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCAM2 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149402
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
Specifications
| NCAM2 | |
| Polyclonal | |
| Unconjugated | |
| NCAM2 | |
| fasciclin II; NCAM2; N-CAM-2; Ncam-2; NCAM21; neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; Ocam; OCAM-GPI; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen; Rncam | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17968 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| O35136 | |
| NCAM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OCAM, Leu20-Gly700. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction