missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCAPG2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550015
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total NCAPG2 protein.
This gene encodes a protein that belongs to the Condensin2nSMC family of proteins. The encoded protein is a regulatory subunit of the condensin II complex which, along with the condensin I complex, plays a role in chromosome assembly and segregation during mitosis. A similar protein in mouse is required for early development of the embryo. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Aug 2013].
Specifications
| NCAPG2 | |
| Polyclonal | |
| Unconjugated | |
| NCAPG2 | |
| 5830426I05Rik; CAPG2; CAP-G2; chromosome-associated protein G2; Condensin-2 complex subunit G2; hCAP-G2; leucine zipper protein 5; LUZP5; mCAP-G2; more than blood homolog; more than blood protein; Mtb; Ncapg2; Non-SMC condensin II complex subunit G2; non-SMC condensin II complex, subunit G2; RGD1308406 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54892 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86XI2 | |
| NCAPG2 | |
| Synthesized peptide derived from internal of human NCAPG2 (aa 561-610). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction