missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAPH2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28723JF646
This item is not returnable.
View return policy
Description
NCAPH2 Polyclonal antibody specifically detects NCAPH2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| NCAPH2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CAP-H2, CAP-H2 subunit of the condensin II complex, CAPH2,384D8-2, Chromosome-associated protein H2, condensin-2 complex subunit H2, hCAP-H2, kleisin beta, Kleisin-beta, MGC15858, MGC18000, MGC2455, MGC4133, MGC5305, MGC8640, Non-SMC condensin II complex subunit H2, non-SMC condensin II complex, subunit H2 | |
| The immunogen recognized by this antibody maps to a region between residue 425 and 475 of human chromosome-associated protein H2 using the numbering given in entry NP_689512.2 (GeneID 29781). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 29781 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction