missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCCRP1 Antibody (5C.6), Novus Biologicals™
Mouse Monoclonal Antibody
€ 195.00
Specifications
| Antigen | NCCRP1 |
|---|---|
| Clone | 5C.6 |
| Concentration | 0.73 mg/mL |
| Dilution | Western Blot 10 μg/mL, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Inhibition Activity |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18531102
|
Novus Biologicals
NB100-2071-50ug |
50 μg | |||||||
Description
NCCRP1 Monoclonal specifically detects NCCRP1 in Rat, Porcine, Bovine, Canine, Equine, Fish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Inhibition Activity.Specifications
| NCCRP1 | |
| 0.73 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Immunology | |
| Q6ZVX7 | |
| 342897 | |
| Purified NCC cells from catfish. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 5C.6 | |
| Western Blot 10 μg/mL, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Inhibition Activity | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| LOC342897, NCCRP-1, nonspecific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog, non-specific cytotoxic cell receptor protein 1 homolog (zebrafish) | |
| NCCRP1 | |
| IgM | |
| Immunogen affinity purified | |
| Nonspecific Cytotoxic Cell (5C.6) |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title