missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCDN Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142252
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_055099.1; NP_001014841.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Probably involved in signal transduction, in the nervous system, via increasing cell surface localization of GRM5 and positively regulating its signaling. Required for the spatial learning process. Acts as a negative regulator of Ca(2+)-calmodulin-dependent protein kinase 2 (CaMK2) phosphorylation. May play a role in modulating melanin-concentrating hormone-mediated functions via its interaction with MCHR1 that interferes with G protein-coupled signal transduction. May be involved in bone metabolism. May also be involved in neurite outgrowth.
Specifications
| NCDN | |
| Polyclonal | |
| Unconjugated | |
| NCDN | |
| AU042419; Kiaa0607; minisatellite 10ae detected by probe MMS10; mKIAA0607; MMS10-AE; Ms10ae; M-Sema F-associating protein of 75 kDa; NCDN; Neurite outgrowth-related protein from the rat brain; neurochondrin; neurochondrin-1; neurochondrin-2; Norbin; Sfap75; similar to neurochondrin | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 23154 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9UBB6 | |
| NCDN | |
| Peptide with sequence C-HYLLQVGSEKQKEP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction