missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCE2 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142723

Product Code. 17868222

  • € 474.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NCE2
Polyclonal
Unconjugated
UBE2F
2510010F15Rik; AI851109; MGC18120; NCE2; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RGD1307608; RING-type E3 NEDD8 transferase UBE2F; UBE2F; Ube2fa; ubiquitin conjugating enzyme E2 F (putative); ubiquitin conjugating enzyme E2F (putative); ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative)
Goat
Ammonium sulfate precipitation
RUO
363284
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q5U203
UBE2F
Peptide with sequence SKLKRDDGLKGSRT-C.
100 μg
Primary
Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.