missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCF4 Antibody (OTI4B5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72893DL594
This item is not returnable.
View return policy
Description
NCF4 Monoclonal antibody specifically detects NCF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| NCF4 | |
| Monoclonal | |
| DyLight 594 | |
| MGC3810, NCF, NCF-4, neutrophil cytosol factor 4, neutrophil cytosolic factor 4 (40kD), neutrophil cytosolic factor 4, 40kDa, Neutrophil NADPH oxidase factor 4, p40phox, p40-phox, SH3 and PX domain-containing protein 4, SH3PXD4P40PHOX | |
| Full length recombinant protein of human NCF4 (NP_000622) produced in HEK293T cells. | |
| 0.1 mL | |
| Signal Transduction | |
| 4689 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction