Learn More
Invitrogen™ NCK2 Recombinant Rabbit Monoclonal Antibody (24GB105)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557100
 View more versions of this product
Product Code. 30275876
  • € 151.00 / 20µL
Description
NCK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCK2
Recombinant Monoclonal
2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O43639
NCK2
A synthesized peptide derived from human Nck2 (aa 50-100).
20 μL
Primary
Human
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
24GB105
Unconjugated
NCK2
4833426I10Rik; cytoplasmic protein NCK2; Grb4; growth factor receptor-bound protein 4; NCK adaptor protein 2; NCK2; Nck-2; NCKbeta; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Rabbit
Affinity chromatography
RUO
8440
-20°C