missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKAP1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26086
This item is not returnable.
View return policy
Description
NCKAP1 Polyclonal specifically detects NCKAP1 in Human, Mouse, Rat, Porcine, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| NCKAP1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| NCKAP1 | |
| Peptide with sequence C-TPREYLTSHLEIR corresponding to internal region according to NP_995314.1. | |
| 0.1 mg | |
| Apoptosis | |
| 10787 | |
| Human, Mouse, Rat, Pig, Bovine, Canine | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:128000 | |
| FLJ11291, HEM2p125Nap1, KIAA0587, Membrane-associated protein HEM-2, MGC8981, NAP 1, NAP125, NAP1Nap1, NCK-associated protein 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_038464.1; NP_995314.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction