Learn More
Novus Biologicals™ NCKAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-29455
Product Code. 30046434
  • € 571.00 / 100µg
Description
NCKAP1 Polyclonal antibody specifically detects NCKAP1 in Human samples. It is validated for Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCKAP1
Polyclonal
Tris-citrate/phosphate pH 7 -8.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation
Unconjugated
FLJ11291, HEM2p125Nap1, KIAA0587, Membrane-associated protein HEM-2, MGC8981, NAP 1, NAP125, NAP1Nap1, NCK-associated protein 1
Between 925 and 975
100 μg
Apoptosis
10787
Store at 4°C. Do not freeze.