missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCKAP1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13239068
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13239068 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13239068 Supplier Invitrogen™ Supplier No. PA521680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: NCKAP1-transfected 293T, Mouse brain, rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Xenopus laevis (96%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene interacts with Nck and may be involved in the downstream signaling of Nck and shares similarity with the mouse Dhm1 and the yeast dhp1 gene. The yeast gene is involved in homologous recombination and RNA metabolism, such as RNA synthesis and RNA trafficking. Complementation studies show that Dhm1 has a similar function in mouse as dhp1. The function of the human gene has not yet been determined. Transcript variants encoding different isoforms have been noted for this gene; however, their full-length nature is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCKAP1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene NCKAP1
Gene Accession No. P28660, P55161, Q9Y2A7
Gene Alias Brain protein H19; C79304; H19; HEM2; Hem-2; KIAA0587; Membrane-associated protein HEM-2; MH19; mKIAA0587; NAP 1; Nap1; NAP125; NCK associated protein 1; NCKAP1; NCK-associated protein 1; p125Nap1
Gene Symbols NCKAP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NCKAP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10787, 50884, 58823
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.