missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKIPSD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19492
This item is not returnable.
View return policy
Description
NCKIPSD Polyclonal specifically detects NCKIPSD in Human, Mouse samples. It is validated for Western Blot.
Specifications
| NCKIPSD | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, AF3p21, AF3P21DIP-1, dia interacting protein, Dia-interacting protein 1, diaphanous protein interacting protein, Diaphanous protein-interacting protein, DIP, DIP1, MGC23891, NCK interacting protein with SH3 domain, ORF1, SPIN90VIP54, WASLBP, WASP-interacting SH3-domain protein, WISHSH3 adapter protein SPIN90, Wisk90 kDa | |
| Rabbit | |
| 79 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (91%), Bovine (96%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.86mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NZQ3 | |
| NCKIPSD | |
| Recombinant protein encompassing a sequence within the C-terminus region of human NCKIPSD. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 51517 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction