missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKIPSD, Mouse anti-Human, Clone: 1A10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051517-M11.100ug
This item is not returnable.
View return policy
Description
Sequence: MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHHSpecifications
| NCKIPSD | |
| Monoclonal | |
| Unconjugated | |
| NCK interacting protein with SH3 domain | |
| AF3P21/DIP/DIP1/MGC23891/ORF1/SPIN90/WASLBP/WISH | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1A10 | |
| In 1x PBS, pH 7.4 | |
| NM_184231 | |
| NCKIPSD | |
| NCKIPSD (NP_909119.1, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51517 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction