missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA2 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-388AF750
This item is not returnable.
View return policy
Description
NCOA2 Polyclonal antibody specifically detects NCOA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| NCOA2 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| BHLHE75, bHLHe75MGC138808, Class E basic helix-loop-helix protein 75, EC 2.3.1.48, GRIP1glucocorticoid receptor-interacting protein-1, KAT13C, NCoA-2SRC2, nuclear receptor coactivator 2, TIF2hTIF2, Transcriptional intermediary factor 2 | |
| The immunogen recognized by this antibody maps to a region between residues 700 and 750 of human Nuclear Receptor Coactivator 2 using the numbering given in entry NP_006531.1 (GeneID 10499). | |
| 0.1 mL | |
| Cancer, DNA Repair, Transcription Factors and Regulators | |
| 10499 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction