missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA3/AIB1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-314B
This item is not returnable.
View return policy
Description
NCOA3/AIB1 Polyclonal antibody specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot
Specifications
| NCOA3/AIB1 | |
| Polyclonal | |
| Western Blot | |
| ACTRCBP-interacting protein, AIB1AIB-1, Amplified in breast cancer 1 protein, BHLHE42, bHLHe42p/CIP, CAGH16, Class E basic helix-loop-helix protein 42, CTG26, KAT13B, MGC141848, NCoA-3, nuclear receptor coactivator 3, pCIP, RAC-3, RAC3pCIP, Receptor-associated coactivator 3, SRC3, SRC-3EC 2.3.1.48, Steroid receptor coactivator protein 3, Thyroid hormone receptor activator molecule 1, TNRC16, TRAM1, TRAM-1TNRC14 | |
| A synthetic peptide, which represented a portion of human nuclear receptor coactivator 3 encoded within exon 13 (LocusLink ID 8202). | |
| 0.1 mL | |
| Breast Cancer, Cancer, Chromatin Research, Transcription Factors and Regulators | |
| 8202 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction