Learn More
NCOA3/AIB1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-314R
Product Code. 18743073
  • € 633.00 / 0.10mL
Description
NCOA3/AIB1 Polyclonal antibody specifically detects NCOA3/AIB1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot
A synthetic peptide, which represented a portion of human nuclear receptor coactivator 3 encoded within exon 13 (LocusLink ID 8202).
Immunogen affinity purified