missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28720
This item is not returnable.
View return policy
Description
NCOA4 Polyclonal specifically detects NCOA4 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| NCOA4 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8031 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot, Immunoprecipitation 10 ug/mg lysate | |
| NP_005428.1 | |
| NCOA4 | |
| The immunogen recognized by this antibody maps to a region between residue 564 and 614 of human 70 kDa AR-activator (nuclear receptor coactivator 4) using the numbering given in entry NP_005428.1 (GeneID 8031). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction