missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA4 Antibody (OTI2G4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46113
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
NCOA4 Monoclonal specifically detects NCOA4 in Human samples. It is validated for Western Blot.
Specifications
| NCOA4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 8031 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| NCOA4 | |
| Human recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4(NP_001138732) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction