missing translation for 'onlineSavingsMsg'
Learn More

NCOA4 Monoclonal Antibody (OTI2G10), TrueMAB™, OriGene

Product Code. p-200019116
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17546653 30 μL 30µL
17526653 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17546653 Supplier Origene Technologies Supplier No. TA806471S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCOA4 Monoclonal antibody specifically detects NCOA4 in Human samples. It is validated for Western Blot

Specifications

Antigen NCOA4
Applications Western Blot
Classification Monoclonal
Clone OTI2G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NCOA4
Gene Accession No. Q13772
Gene Alias 70 kDa androgen receptor coactivator; 70 kDa AR-activator; Androgen receptor coactivator 70 kDa protein; Androgen receptor-associated protein of 70 kDa; ARA70; ELE1; NCOA4; NCoA-4; Nuclear receptor coactivator 4; PTC3; ret fused; RET-activating gene ELE1; Ret-activating protein ELE1; RFG
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8031
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.