missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA6 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-336DL594
This item is not returnable.
View return policy
Description
NCOA6 Polyclonal antibody specifically detects NCOA6 in Human samples. It is validated for Western Blot
Specifications
| Western Blot | |
| DyLight 594 | |
| The immunogen recognized by this antibody maps to a region between residue 2000 and the C-terminus (residue 2063) of human Activating Signal Cointegrator-2 using the numbering given in Swiss-Prot entry Q14686 (GeneID 23054). | |
| 23054 |
| Polyclonal | |
| Activating signal cointegrator 2, AIB3NRC RAP250, Amplified in breast cancer protein 3, ASC-2, Cancer-amplified transcriptional coactivator ASC-2, KIAA0181PPAR-interacting protein, NRCamplified in breast cancer-3 protein, nuclear receptor coactivator 6, Nuclear receptor coactivator RAP250, Nuclear receptor-activating protein, 250 kDa, peroxisome proliferator-activated receptor interacting protein, PRIPPeroxisome proliferator-activated receptor-interacting protein, RAP250Thyroid hormone receptor-binding protein, thyroid hormone receptor binding protein, TRBPASC2activating signal cointegrator-2 | |
| Immunogen affinity purified | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction