missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25241-1-AP-20UL
This item is not returnable.
View return policy
Description
Nuclear receptor coactivator that directly binds nuclear receptors and stimulates the transcriptional activities in a hormone-dependent fashion. Coactivates expression in an agonist- and AF2-dependent manner. Involved in the coactivation of different nuclear receptors, such as for steroids (GR and ERs), retinoids (RARs and RXRs), thyroid hormone (TRs), vitamin D3 (VDR) and prostanoids (PPARs). Probably functions as a general coactivator, rather than just a nuclear receptor coactivator. May also be involved in the coactivation of the NF-kappa-B pathway. May coactivate expression via a remodeling of chromatin and its interaction with histone acetyltransferase proteins.Specifications
| NCOA6 | |
| Polyclonal | |
| Unconjugated | |
| NCOA6 | |
| AIB3, ANTP, ASC 2, ASC2, HOX1.1, HOXA7, KIAA0181, NCOA6, NRC, NRC RAP250, nuclear receptor coactivator 6, PPAR interacting protein, PRIP, RAP250, TRBP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23054, 56406 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14686, Q9JL19 | |
| Ncoa6 | |
| NCOA6 Fusion Protein Ag18737 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction