missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85199
This item is not returnable.
View return policy
Description
NCOA7 Polyclonal specifically detects NCOA7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NCOA7 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| 140 kDa estrogen receptor-associated protein, dJ187J11.3, ERAP140Nbla00052, ESNA1, Estrogen nuclear receptor coactivator 1, estrogen receptor associated protein 140 kDa, FLJ45605, MGC88425, Nbla10993, nuclear receptor coactivator 7, putative protein product of Nbla00052, putative protein product of Nbla10993 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 135112 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| NCOA7 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EPLPVKLNSSTEANVIKEALDSSLESTLDNSCQGAQMDNKSEVQLWLLKRIQVPIEDILPSKEEKSKTPPMFLCIKVGKPMRKSFATHTAAMVQQYGKRR | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction