Learn More
Invitrogen™ NCOA7 Monoclonal Antibody (OTI1E12)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527306
This item is not returnable.
View return policy
Description
NCOA7 Monoclonal Antibody for Western Blot
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
Specifications
| NCOA7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NI08 | |
| NCOA7 | |
| Full length human recombinant protein of NCOA7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E12 | |
| Unconjugated | |
| NCOA7 | |
| 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 135112 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |