Learn More
Invitrogen™ NCOA7 Monoclonal Antibody (OTI1E12)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527306
Product Code. 15966894
  • € 572.00 / 100µL
Description
NCOA7 Monoclonal Antibody for Western Blot
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NCOA7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NI08
NCOA7
Full length human recombinant protein of NCOA7 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1E12
Unconjugated
NCOA7
140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4
Mouse
Affinity Chromatography
RUO
135112
-20°C, Avoid Freeze/Thaw Cycles