missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA7 Monoclonal Antibody (OTI1E12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810478
This item is not returnable.
View return policy
Description
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.Specifications
| NCOA7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NI08 | |
| NCOA7 | |
| Full length human recombit protein of human NCOA7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1E12 | |
| Unconjugated | |
| NCOA7 | |
| ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 135112 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction