missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCOA7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15744341 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15744341 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15744341 Supplier Invitrogen™ Supplier No. PA556431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EPLPVKLNSS TEANVIKEAL DSSLESTLDN SCQGAQMDNK SEVQLWLLKR IQVPIEDILP SKEEKSKTPP MFLCIKVGKP MRKSFATHTA AMVQQYGKRR Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCOA7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCOA7
Gene Accession No. Q8NI08
Gene Alias 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4
Gene Symbols NCOA7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NCOA7. Recombinant protein control fragment (Product #RP-94663).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.