missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NCOA7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575420
This item is not returnable.
View return policy
Descripción
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
Especificaciones
| NCOA7 | |
| Polyclonal | |
| Unconjugated | |
| NCOA7 | |
| 140 kDa estrogen receptor-associated protein; 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nbla00052; Nbla10993; Ncoa7; NCOA7-AS; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; RGD1566426; TBC/LysM-associated domain containing 4; TLDC4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 135112, 211329, 498995 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.2 | |
| Q6DFV7, Q8NI08 | |
| NCOA7 | |
| Synthetic peptide corresponding to amino acids 422-470 of Human NCOA7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido