missing translation for 'onlineSavingsMsg'
Learn More

ND1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16859184 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16859184 20 μL 20µL
16849184 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16859184 Supplier Proteintech Supplier No. 197031AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects both isoforms of ND1, 318aa 34-36 kDa and 268aa, 28-30 kDa.

A small number of complex I subunits are the products of mitochondrial genes (subunits 1-7),while the remainder are nuclear encoded and imported from the cytoplasm. NADH dehydrogenase subunit 1 (ND1) binds rotenone and rotenone analogs and might be involved in electron transfer to ubiquinone. Mutations in the ND1 gene may be implicated in several disorders, including Leber hereditary optic neuropathy, Alzheimer′s disease and Parkinson disease.

Specifications

Antigen ND1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MT-ND1
Gene Accession No. P03886
Gene Alias MT ND1, MTND1, NADH dehydrogenase subunit 1, NADH1, ND1
Gene Symbols mt-Nd1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17716, 4535
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.