missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ND1-YL2

Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
ND1-YL2 is a peptide-based PROTAC™ Degrader of steroid receptor co-activator 1 (SRC-1; also known as nuclear receptor coactivator 1, NCOA1). ND1-YL2 is composed of a stapled peptide that binds SRC-1 (YL2) joined by a linker to a tetrapeptide that binds UBR box domains. Upon ternary complex formation, SRC-1 is polyubiquitinated and subsequently degraded via the N-degron pathway. This Degrader induces dose-dependent degradation of SRC-1 in the MDA-MB-231 triple negative breast cancer cell line (DC50 = 10 μM), and binds to the PAS-B domain of SRC-1 (Ki = 320 nM). ND1-YL2 inhibits MDA-MB-231 cell migration in vitro, and suppresses metastasis of MDA-MB-231 cells in vivo.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Purity | 95% |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | M.W.-observed: 2385.93 |
| Quantity | 100 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction